Learn More
Tocris Bioscience™ MRS 5698

Description
Specifications
Specifications
| Purity | >96% |
| Quantity | 1 mg |
| Product Type | MRS 5698 |
| Molecular Weight (g/mol) | 564.97 |
| Inhibitors | MRS 5698 |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.