Learn More
Medchemexpress LLC MRT-2359 10 MG
SDP
Supplier:  Medchemexpress LLC HY15335610MG
Encompass_Preferred
MRT-2359 / CAS No. 2803881-11-8 / Molecular Weight- 495.38MRT-2359 is an orally active and potent degrader of GSPT1 with anti-tumor activity. MRT-2359 also inhibits the growth of resistant non-small cell lung cancer /NSCLC/ and small cell lung cancer /SCLC/ cells. For research use only.
Catalog No. NC2319740
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.