Learn More
Apexbio Technology LLC MRT67307 HCl, 50mg. Cas: 1190378-57-4 (free-base) MFCD: MFCD19443643. SIK/TBK-1/IKKe inhibitor
SDP
Supplier:  Apexbio Technology LLC A363550
Encompass_Preferred
MRT67307 is an inhibitor for TBK1, IKK , MARK1-4 and NUAK1 with IC50 value of 19, 160, 27-52 and 230nM , respectively [1]. It is an inhibitor for ULK1and ULK2 with IC50 value of 45 and 38nM, respectively [2]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4820
Due to product restrictions, please to purchase or view availability for this product.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.