Learn More
Selleck Chemical LLC MRTX-1257 S8831-5MG
SDP
Supplier:  Selleck Chemical LLC S88315MG
Encompass_Preferred
MRTX-1257 is a potent selective irreversible covalent and orally active inhibitor of KRAS G12C with IC50 of 900 pM for KRAS dependent ERK phosphorylation in H358 cells
Catalog No. 50-217-3397
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.