Learn More
TARGETMOL CHEMICALS INC MRTX1133 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T9303
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MRTX1133 is a potent selective and noncovalent inhibitor of KRAS G12D. MRTX1133 exhibits an estimated KD against KRAS G12D of 0.2 pM. MRTX1133 inhibits mutant KRAS-dependent signal transduction by preventing SOS1-catalyzed nucleotide exchange and/or formation of the KRAS G12D/GTP/RAF1 complex. MRTX1133 selectively inhibits KRAS G12D mutant tumor cells with no effect on KRAS wild-type. MRTX1133 exhibits single digit nanomolar activity in cellular assays and marked in vivo efficacy in tumor models harboring KRAS G12D mutations. purity: 99%
Catalog No. 50-257-9010
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.