Learn More
REPLIGEN CORPORATION Repligen MiniKros Sampler, 20 cm Length, 0.2 µm PES Membrane, 3/4" TC Connections
SDP
Supplier:  REPLIGEN CORPORATION S02P20U10N
Encompass
The MiniKros Sampler is designed for precise tangential flow filtration in scientific applications. Its 0.2 µm PES membrane effectively clarifies and concentrates biological samples, ensuring reliable performance in bioprocessing. With 3/4" TC connections, it guarantees secure integration into systems while maintaining aseptic conditions. This module stands out for its optimal design tailored for demanding filtration processes, differentiating it from typical filters with less precision.
Catalog No. NC1896812
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.