Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ MS 21570
GREENER_CHOICE

Catalog No. p-7116119
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
62-981-0 10 mg
62-985-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 62-981-0 Supplier Tocris Bioscience™ Supplier No. 6298/10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

GPR171 antagonist

MS 21570 is a GPR171 antagonist (IC50 = 220 nM). Inhibits BigLEN (Cat.No. 6304) mediated increases in [35S]GTPγS binding in hypothalamic membranes and hyperpolarization of basolateral amygdala pyramidal neurons. Reduces anxiety-like behavior and fear conditioning in mice.

Specifications

Chemical Name or Material N-Methyl-5-[(phenylmethyl)thio]-1,3,4-thiazadiazol-2-amine
CAS 65373-29-7
Assay Percent Range >98%
Quantity 10 mg
Molecular Formula C10H11N3S2
Formula Weight Observed MW: 237.34
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.