Learn More
Biotium EGFRvIII (Epidermal Growth Factor Receptor, Variant III) (GFR/2600R), 0.2mg/mL: (100uL)
Supplier: Biotium BNUB2600100
EGFR is type I receptor tyrosine kinase with sequence homology to erbB-1, -2, -3 -4 or HER-1, -2, -3 -4. It binds to Epidermal Growth Factor (EGF), Transforming Growth Factor-a (TGF-a), Heparin-binding EGF (HB-EGF), amphiregulin, betacellulin and epiregulin. EGFR is overexpressed in tumors of breast, brain, bladder, lung, gastric, head & neck, esophagus, cervix, vulva, ovary, and endometrium. It is predominantly present in squamous cell carcinomas. Supplied as 100 uL of 0.2 mg/mL purified antibody with 0.05% BSA and 0.05% sodium azide in PBS.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.