Learn More
Promega MS Compatible Human Protein Extract, Intact
Catalog No. PRV6941 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV6941 1 mg
1 options
Catalog No. PRV6941 Supplier Promega Supplier No. V6941
Mass spectrometry-compatible yeast and human protein extracts are designed specifically for mass spectrometry applications. The extracts are predigested and cleaned up by solid-phase extraction for immediate use in liquid chromatography/mass spec analysis
  • Protein Extracts for Easier Mass Spec Instrument Monitoring
  • Choose human or yeast, digested or intact extract
  • Monitor LC and MS instrument performance
  • Use for method development and optimization
Specifications
Content And Storage <-65°C
Quantity 1 mg