Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spi-B Rat anti-Mouse, PE, Clone: U14-267, BD

Catalog No. bdb565834 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB565834 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB565834 Supplier BD Biosciences Supplier No. 565834

Rat Monoclonal Antibody

The U14-267 monoclonal antibody specifically recognizes mouse Spi-B, that is encoded by Spib (Spi-B transcription factor (Spi-1/PU.1 related)). Spi-B belongs to the Ets family of transcription factors. It is expressed predominantly in B cells, T-cell progenitors, and plasmacytoid dendritic cells. Spi-B plays crucial roles in the development of T cells and plasmacytoid dendritic cells, and in the terminal differentiation of B cells. Although Spi-B deficient mice have normal numbers of B cells, they exhibit defects in germinal center formation and maintenance. In contrast, overexpression of Spi-B blocks plasma cell differentiation due to its capacity to repress Blimp-1 and Xbp-1 expression.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Spi-B
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone U14-267
Concentration 0.2mg/mL
Conjugate PE
Description SPI-B; Spib; Spi-1/PU.1 related; Ets transcription factor Spi-B
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse Spi-B Recombinant Protein
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.