Learn More
Waters Corp MS UPLC Columns Method Development Kit 2.1 mm x 50 mm, 4/pk
Supplier:  Waters Corp 176001607
Encompass_Preferred
The MS UPLC method development kit combines 1.7 µm BEH C18, with 1.8 µm HSS C18, C18 SB and T3 column chemistries to accomodate your method development approach, enabling methods to be developed efficiently and effectively. By utilizing UPLC Technology for method development, a substantial improvement in throughput can be realized. This, in turn, reduces cost per sample and time of analysis considerably while maintaining or improving separation integrity, enabling products to be brought to market faster and improving the overall profitability of the assay.
Catalog No. 50-791-021
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.