Learn More
TARGETMOL CHEMICALS INC MS049 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T437825MG
Encompass_Preferred
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. MS 049 is a potent, selective, and cell-active dual inhibitor of PRMT4 and PRMT6 with IC 50 of 34 nM and 43 nM, respectively. Purity 98%
Catalog No. 50-287-8669
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.