Learn More
Medchemexpress LLC HY-133129 5mg Medchemexpress, MS1943 CAS:2225938-17-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1331295MG
Encompass_Preferred
Medchemexpress, HY-133129 5mg MS1943 CAS:2225938-17-8 MS1943 is a first-in-class, orally bioavailable EZH2 selective degrader, with an IC50 of 120 nM. MS1943 significantly reduces EZH2 protein levels in numerous triple-negative breast cancer (TNBC) and other cancer and noncancerous cell lines. MS1943 effectively blocks proliferation of multiple TNBC and other cancer cell lines. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0283
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.