Learn More
Invitrogen™ MS4A14 Polyclonal Antibody

Description
MS4A14 Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | MS4A14 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | MS4A14 |
| Gene Accession No. | Q96JA4 |
| Gene Alias | membrane spanning 4-domains A14; membrane-spanning 4-domains subfamily A member 14; membrane-spanning 4-domains, subfamily A, member 14; membrane-spanning 4-domains, subfamily A, member 16; MS4A13; MS4A13 protein; MS4A14; MS4A16; NYD-SP21; testes development-related NYD-SP21; testis development protein NYD-SP21 |
| Gene Symbols | MS4A14 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.