Learn More
Invitrogen™ MS4A4A Recombinant Mouse Monoclonal Antibody (3F2)

Description
Specificity: This antibody is specific for human extracellular MS4A4A.
Specifications
Specifications
| Antigen | MS4A4A |
| Applications | Flow Cytometry, Immunoprecipitation |
| Classification | Recombinant Monoclonal |
| Clone | 3F2 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% ProClin 300 |
| Gene | MS4A4A |
| Gene Accession No. | Q96JQ5 |
| Gene Alias | 4SPAN1; CD20 antigen-like 1; CD20L1; CD20-L1; Fc epsilon receptor beta subunit homolog; four-span transmembrane protein 1; HDCME31P; membrane spanning 4-domains A4A; membrane-spanning 4-domains subfamily A member 4A; membrane-spanning 4-domains, subfamily A, member 4; membrane-spanning 4-domains, subfamily A, member 4A; MS4A4; MS4A4A; MS4A7 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.