Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MS4A6A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572732 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572732 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572732 Supplier Invitrogen™ Supplier No. PA572732
  • $586.00
    $478.65 / Each of 1

    Save $107.35

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

MS4A6A Polyclonal Antibody for Western Blot, IHC (P), ELISA

May be involved in signal transduction as a component of a multimeric receptor complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MS4A6A
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene MS4A6A
Gene Accession No. Q9H2W1
Gene Alias 4SPAN3; 4SPAN3.2; CD20 antigen-like 3; CD20L3; CD20-like precusor; CDA01; four-span transmembrane protein 3; four-span transmembrane protein 3.1; four-span transmembrane protein 3.2; HAIRB-iso; membrane spanning 4-domains A6A; membrane-spanning 4-domains subfamily A member 6A; membrane-spanning 4-domains subfamily A member 6D; membrane-spanning 4-domains, subfamily A, member 11; membrane-spanning 4-domains, subfamily A, member 6A; membrane-spanning 4-domains, subfamily A, member 6A, isoform 2; Ms4a11; MS4A6; MS4A6A; MS4A6A-polymorph; MST090; MSTP090
Gene Symbols MS4A6A
Host Species Rabbit
Immunogen MS4A6A antibody for a 19 amino acid synthetic peptide near the amino terminus of human MS4A6A. The immunogen is within the first 50 amino acids of MS4A6A.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 361735, 64231
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.