Learn More
MSA™ Abrasi-Blast™ Supplied-Air Respirator Lens Cartridges
Catalog No. 19047241 Shop All MSA Products
Change view
Click to view available options
For Use With (Application):
Heavy blasting
Medium blasting
Thickness:
0.09 in.
0.12 in.
Capacity (English):
2/Carton
3/Carton
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application) Thickness Capacity (English)
19-047-241 Medium blasting 0.09 in. 3/Carton
18-999-3215 Heavy blasting 0.12 in. 2/Carton
18-999-3181 Heavy blasting 0.12 in. 3/Carton
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-047-241 Supplier MSA™ Supplier No. 473798
Add to Cart
Add to Cart
Cover lens cartridge assembly to match different blasting levels.
  • Cover lens cartridge assembly for Abrasi-Blast respirators
  • Removable lenses feature tabs which are pulled to remove cover lenses as abrasion occurs
  • Used lenses can be stored in the hood front pocket
  • Different lens cartridge types are available to match different blasting levels
Specifications
Capacity (English) 3/Carton
Certifications/Compliance NIOSH
Material Untempered
Product Type Lens Cartridge
For Use With (Application) Medium blasting
Lens Type Untempered
Model 473798
Quantity 12/Pk.
Thickness 0.09 in.