Learn More
MSA™ Accessories for Air-Supplied Hood
Catalog No. 189993640 Shop All MSA Products
Change view
Click to view available options
Socket
Catalog No.
18-999-3640 Socket
1 options
Catalog No. 18-999-3640 Supplier MSA™ Supplier No. 484236
Add to Cart
Add to Cart
Snap-tite female socket for use with air-supplied hood.
  • Accessory for SAR hood
  • Snap-tite female socket
  • Connects 0.50 in. hoses to manifold and air source
Specifications
Certifications/Compliance NIOSH, MSHA
Product Type Hose Socket
For Use With (Application) Connect air supply hoses to manifold and air source
For Use With (Equipment) Quick-disconnects of Versa-hood Supplied Air Respirator Hood
Includes 0.37 in. MPT adapter
Model 484236
Quantity 1 Ea.