Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ Advantage™ 4000 Full Facepiece Respirators, Adapter Not Included

Catalog No. p-4246860 Shop All MSA Products
Change view
Click to view available options
Size:
Small
Medium
Large
Material:
Hycar™
Silicone
Harness Material:
Polyester Net Head Harness
Rubber
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Material Harness Material
18-139-413 Small Hycar™ Polyester Net Head Harness
18-139-415 Small Hycar™ Rubber
18-139-414 Small Silicone Rubber
18-139-418 Medium Silicone Rubber
18-139-417 Medium Hycar™ Polyester Net Head Harness
18-139-419 Medium Hycar™ Rubber
18-139-421 Large Hycar™ Polyester Net Head Harness
18-139-423 Large Hycar™ Rubber
18-139-422 Large Silicone Rubber
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 18-139-413 Supplier MSA™ Supplier No. 10083797
Only null left
Add to Cart
Add to Cart

Full-face reusable respirators are ideal for a variety of applications. Advantage™ 4000 series is compatible with several MSA respirator models, including air-purifying respirators, supplied-air respirators and powered air-purifying respirators. Adapter NOT included.

  • Single-port facepiece
  • Compatible with MSA air-purifying respirators, supplied-air respirators and powered air-purifying respirators
  • NIOSH-approval 42 CFR Part 84
  • 4-pt suspension with rubber or polyester net head harness
  • Standand fog-reducing nosecup
  • Facepiece material is available in silicone or Hycar™
  • Three standard sizes to ensure a secure fit
  • Components lock into place in seconds for quick, convenient performance
  • Ideal for use in utilities, mining, oil and gas, fire service, construction, confined space, asbestos abatement, welding and sanding/grinding applications

Specifications

ANSI Standard ANSI Z87.1 2003
Certifications (NIOSH) NIOSH 42 CFR Part 84
Color Black
Material Hycar™
Product Type Full Facepiece Respirator
Size Small
Facepiece Material Hycar™
For Use With (Application) Utilities, mining, oil and gas, fire service, construction, confined space
For Use With (Equipment) Comfo series aerosol filters and chemical cartridges
Harness Material Polyester Net Head Harness
Material (Coating) Hard coating
Material (Lens) Polycarbonate
Model 10083797
No. of Ports Single
Suspension Type 4-point
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.