Learn More
MSA™ Affinity™ Pro Sizing and Fit Check Cups
Catalog No. 189993984 Shop All MSA Products
Change view
Click to view available options
Kit Fit Sizing Cup S/m/lg 3/pk
Catalog No.
18-999-3984 Kit Fit Sizing Cup S/m/lg 3/pk
1 options
Catalog No. 18-999-3984 Supplier MSA™ Supplier No. 816659
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
For Affinity Pro respirators. Fit check cup allows the wearer to perform a face-seal check prior to use to provide optimal respirator protection. Sizing cup ensures that the correct size respirator is selected.
Order Info
Sizing cups supplied in packs of three; each pack includes one small, one medium, and one large size.
Specifications
Model 816659