Learn More
MSA™ Air Cylinders
Catalog No. 19121553 Shop All MSA Products
Change view
Click to view available options
Cylinder; Valve assembly
Catalog No.
19-121-553 Cylinder; Valve assembly
1 options
Catalog No. 19-121-553 Supplier MSA™ Supplier No. 816115
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
30 minute-rated air supply cylinder for lighter weight maneuverability
Carbon fibers have a high strength-to-weight ratio
  • A final over-wrap of fiberglass fibers enhances abrasion resistance
  • A glow-in-the-dark luminescent band is applied for improved night visibility
Firefighting
Specifications
Product Type Cylinder Valve Assembly