Learn More
MSA™ AirHawk™ SCBA Stock Configurations
Catalog No. 19815611 Shop All MSA Products
Change view
Click to view available options
Product Type:
Airhawk SCBA with hard case
Airhawk SCBA with no case
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
19-815-611 Airhawk SCBA with no case
19-815-610 Airhawk SCBA with hard case
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-815-611 Supplier MSA™ Supplier No. 10043202
Popular configurations
For detailed specifications, consult general description for Airhawk SCBAs or contact your Fisher Safety Customer Service Representative.
  • SCBA Respirator, 2216
  • Harness: Nylon with chest strap
  • Facepiece: Ultra Elite, medium, Hycar, rubber head harness
  • Cylinder: Aluminum (30 minutes)
Specifications
Product Type Airhawk SCBA with no case
Duration 30 min.
Pressure 2216 psig
Harness Style Nylon with chest strap, rubber
Material Medium Hycar™