Learn More
MSA™ BIOSENSOR™ Cartridge Plans
Catalog No. 18808922 Shop All MSA Products
Change view
Click to view available options
Product Type:
Cartridge Starter Kit
High Volume Cartridge Kit
Low Volume Cartridge Kit
Medium Volume Cartridge Kit
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
18-808-922 Low Volume Cartridge Kit
18-808-924 High Volume Cartridge Kit
18-808-921 Cartridge Starter Kit
18-808-923 Medium Volume Cartridge Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-808-922 Supplier MSA™ Supplier No. 10084753
Includes
Each biohazard cartridge test kit includes a dry sample kit.
For use with BIOSENSOR 220R Biological Agent Detector Kit
Specifications
Product Type Low Volume Cartridge Kit
For Use With (Equipment) Biosensor™ biological agent detector
Includes 6 Anthrax/Ricin dual kits, 2 Anthrax, 2 Ricin, 6 positive controls, 2 negative control cartridges, 2 wet sample kits