Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ Bitrex™ Qualitative Fit-Test Kit

Catalog No. 19037720 Shop All MSA Products
Change view
Click to view available options
Product Type:
Replacement Fit Test Solution
Replacement Hood
Sensitivity Solution
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
19-037-720 Replacement Hood
19-037-721 Sensitivity Solution
19-037-723 Replacement Fit Test Solution
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-037-720 Supplier MSA™ Supplier No. 697448
Add to Cart
Add to Cart

Includes

Two nebulizer bulbs, testing solution insert, sensitivity test solution insert, test hood, one replacement set of testing and sensitivity nebulizer inserts, and Using the Qualitative Fit-Test Kit videotape.

Safe, efficient, and reliable, this kit has everything necessary to perform OSHA-mandated fit testing on all NIOSH-approved air-purifying respirators.

Bitrex solution contains the chemical compound denatonium benzoate, making it an extremely bitter solution easily detected by the test subject.

Specifications

Product Type Replacement Hood
Certifications/Compliance OSHA, NIOSH
For Use With (Equipment) MSA Bitrex and Saccharin qualitative fit test kit
Model 697448
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.