Learn More
MSA™ Bitrex™ Qualitative Fit-Test Kit
Catalog No. 19035414 Shop All MSA Products
Change view
Click to view available options
Qualitative fit test kit; Universal
Catalog No.
19-035-414 Qualitative fit test kit; Universal
1 options
This item is not returnable. View return policy
Catalog No. 19-035-414 Supplier MSA™ Supplier No. 697444
This item is not returnable. View return policy
Includes
Two nebulizer bulbs, testing solution insert, sensitivity test solution insert, test hood, one replacement set of testing and sensitivity nebulizer inserts, and Using the Qualitative Fit-Test Kit videotape.
Safe, efficient, and reliable, this kit has everything necessary to perform OSHA-mandated fit testing on all NIOSH-approved air-purifying respirators
Bitrex solution contains the chemical compound denatonium benzoate, making it an extremely bitter solution easily detected by the test subject.
Specifications
Product Type Bitrex Fit Test Kit
For Use With (Application) Respirator Fit Testing
Includes Two nebulizer bulbs, testing solution insert, sensitivity test solution insert, test hood, one replacement set of testing and sensitivity nebulizer inserts, and “using the qualitative fit-test kit” videotape
Model 697444
Quantity 1 Ea.