Learn More
MSA™ Bump Cap
Catalog No. 19005370 Shop All MSA Products
Change view
Click to view available options
Color:
Blue
Dark Green
Orange
Royal Blue
White
Yellow
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-005-370 Dark Green
19-005-365 Blue
19-005-366 Yellow
19-005-367 White
19-005-369 Orange
19-005-359 Royal Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19-005-370 Supplier MSA™ Supplier No. 10033655
Add to Cart
Add to Cart
Protect your workers from minor bumps and lacerations
  • Size-adjustable; no replaceable parts
  • Ideal for workplaces such as meat packing, food processing, pest control, meter reading, and automotive repair
  • Perforated sides improves air circulation by allowing cross ventilation
  • Interior plastic liner includes foam pad for additional comfort
  • Standard version is molded from durable high-density polyethylene and is adjustable from size 6.5 to 8
NOTE: Bump caps do not meet the ANSI Z89.1-1997 standard, and are not designed to protect against impact of falling or glancing objects.
Specifications
Color Dark Green
Size Standard
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.