Learn More
MSA™ Cable Slings
Catalog No. 191600208 Shop All MSA Products
Change view
Click to view available options
Certifications/Compliance:
OSHA, ANSI Z359.1-2007, ANSI A10.32
Catalog No. Certifications/Compliance
19-160-0208 OSHA, ANSI Z359.1-2007, ANSI A10.32
1 options
Catalog No. 19-160-0208 Supplier MSA™ Supplier No. SFP3267506
Add to Cart
Add to Cart
Vinyl-coated slings for use in agriculture, asbestos abatement, chemical industry, construction
  • 0.63cm (0.25 in.) galvanized aircraft cable
  • Steel thimble
  • Flemish eye terminations
  • Minimum breaking strength 5,000 lbs
  • Intended to be used in a basket configuration only
  • Cable Sling is approved for use by one person at a time with approved systems
Specifications
Certifications/Compliance OSHA, ANSI Z359.1-2007, ANSI A10.32
For Use With (Equipment) Fall arrest and confined space entry/retrieval component
Length (Metric) 182.8 cm
Product Type Anchorage cable sling
Strength 5000 lbf.
Length (English) 72 in.
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.