Learn More
MSA™ Cascade System Accessory, Dual Gauge Pressure Regulator
Catalog No. 18139766 Shop All MSA Products
Change view
Click to view available options
For Use With (Equipment):
Cascade System
Catalog No. For Use With (Equipment)
18-139-766 Cascade System
1 options
Catalog No. 18-139-766 Supplier MSA™ Supplier No. 68858
Add to Cart
Add to Cart
Dual gauge pressure regulator is used to assemble a 2400 psi cascade system.
Cascade system consists of a bank of respirable-air cylinders that supply a flow of air to dual-purpose air masks or other pressure-demand air-supplied respirators.
Specifications
For Use With (Equipment) Cascade System
Model 68858
Pressure 0 to 3000 psi
Product Type Dual Gauge Pressure Regulator