Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ CBRN and Riot Control Gas Mask Black Cap

Catalog No. 19037424 Shop All MSA Products
Change view
Click to view available options
Certifications/Compliance:
NIOSH
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance
19-037-424 NIOSH
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-037-424 Supplier MSA™ Supplier No. 633297
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Accessory for CBRN and riot control gas mask.

For use with CBRN and riot control gas mask

Specifications

Certifications/Compliance NIOSH
Color Black
Material Acetate
Product Type Black Cap
For Use With (Application) Riot control
For Use With (Equipment) Riot Control Gas Masks
Model 633297
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.