Learn More
MSA™ SCBA Accessory, ClearCommand Voice Amplifier Kit
Catalog No. 19003376 Shop All MSA Products
Change view
Click to view available options
For Use With (Equipment):
SCBA
Catalog No. For Use With (Equipment)
19-003-376 SCBA
1 options
Catalog No. 19-003-376 Supplier MSA™ Supplier No. 10024074
Add to Cart
Add to Cart
Communication system accessory delivers over twice the volume as a facepiece alone.
Enables rescue personnel to communicate clearly over commercially available radios
  • Provides optimal speech communication
  • Robust yet lightweight
  • Works with or without a mask
  • Does not block vision or mechanical communication
  • Features a removable/transferable, water-resistant amplifier per MIL-810E (blowing rain) and submersible internal microphone
  • Attach it to a full facemask (open mode) or put it inside a protective suit (covered mode)
Certifications
Meets DIN 40050-IP54 standards
Specifications
Certifications/Compliance NIOSH,NFPA 1981, 1997, UL Class I, II, III, Division 1, Groups A, B, C, D, E, F and G per ANSI UL 913, Canadian C 22.2 no. 157-92 1987
Applications Firefighting
For Use With (Equipment) SCBA
Includes Bracket and voicemitter
Model 10024074
Product Type Voice Amplifier