Learn More
MSA™ Comfo Classic Probed Respirator Facepieces
Catalog No. 189993676 Shop All MSA Products
Change view
Click to view available options
Size:
Small
Medium
Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-999-3676 Small
18-999-3674 Medium
18-999-3675 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 18-999-3676 Supplier MSA™ Supplier No. 808258
Add to Cart
Add to Cart
This item is not returnable. View return policy
Specifications
Product Type Probed Respirator Facepiece
For Use With (Application) Asbestos abatement, painting, sanding, grinding, welding
Includes Built-in probe assembly, solid brass probe and brass fitting
Material Silicone
Model 808258
Size Small