Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ Comfo™ Classic Respirator Yoke Assembly

Catalog No. 19042392 Shop All MSA Products
Change view
Click to view available options
Size:
Medium
Large
Color:
Gold
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Color
19-042-392 Medium Gold
19-034-092 Large -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-042-392 Supplier MSA™ Supplier No. 807605
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use with Comfo Classic respirators.

  • Accessory for Comfo Classic respirator
  • Available in medium or large sizes

Specifications

Certifications/Compliance NIOSH
Color Gold
Product Type Yoke Assembly
Size Medium
For Use With (Equipment) Comfo Classic respirators
Model 807605
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.