MSA™ Comfo™ and ComfoClassic™ Respirators

Unique face seal design that gives exceptional fit

Manufacturer: MSA™ 808057

 View more versions of this product

Catalog No. 18-999-2589A

  Check Availability
Add to Cart

Required Accessories : Comfo Series cartridges and filters (see separate listings).

    Specifications

    Certifications NIOSH 42CFR84
    Color Black
    Material SoftFeel Hycar
    Model Comfo Classic
    For Use With (Application) Asbestos abatement, painting, sanding, grinding, welding
    Product Type Half-Mask Respirator
    Size Small

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit