Learn More
MSA™ CABA Accessory, Machine Screw
Catalog No. 189994156 Shop All MSA Products
Change view
Click to view available options
Model:
636495
Catalog No. Model
18-999-4156 636495
1 options
Catalog No. 18-999-4156 Supplier MSA™ Supplier No. 636495
Add to Cart
Add to Cart
Compressed airline breathing apparatus (CABA) corresponding accessory provides a supply of safe air wherever air is irrespirable or oxygen deficient.
  • Accessory for compressed airline breathing apparatus (CABA)
  • Allows for independent breathing in irrespirable conditions, including contaminated ambient air and oxygen deficiency
  • Suitable for long-duration work and special applications
  • Modular design for multiple configurations
  • ASV (automatic switch valve) supplies air even if the airline is disconnected or the external air source fails
Specifications
For Use With (Equipment) Compressed Airline Breathing Apparatus
Model 636495
Product Type Screw