Learn More
MSA™ Constant Flow Air-Line Respirators
Catalog No. 189993145 Shop All MSA Products
Change view
Click to view available options
Includes:
Black medium hycar™ ultravue™ full facepiece, hose, web support belt, snap-tite™ aluminum quick-disconnect and high-pressure control valve
Breathing tube, web support belt and air flow control valve
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
18-999-3145 Black medium hycar™ ultravue™ full facepiece, hose, web support belt, snap-tite™ aluminum quick-disconnect and high-pressure control valve
18-999-3175 Breathing tube, web support belt and air flow control valve
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-999-3145 Supplier MSA™ Supplier No. 460863
Specifications
Certifications/Compliance NIOSH, MSHA
Product Type Respirator Assembly
For Use With (Application) Non-IDLH environments
For Use With (Equipment) Constant flow airline respirator
Includes Black medium hycar™ ultravue™ full facepiece, hose, web support belt, snap-tite™ aluminum quick-disconnect and high-pressure control valve
Quantity 1 Ea.