Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ Cover Lens for OptimAir™ 6HC PAPR

Catalog No. 189993635 Shop All MSA Products
Change view
Click to view available options
For Use With (Equipment):
For use with MSA OptimAir 6HC PAPR
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
18-999-3635 For use with MSA OptimAir 6HC PAPR
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-999-3635 Supplier MSA™ Supplier No. 456975
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Color Clear
Product Type Lens Cover
For Use With (Equipment) For use with MSA OptimAir 6HC PAPR
For Use With (Application) Protects facepiece lens from scratches during storage, handling and use
Length (English) 8 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.