Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ Cylinder Valve Burst Disc and Gasket Kit, 3000 PSI

Catalog No. MSA494928 Shop All MSA Products
Change view
Click to view available options
Model:
494928
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
MSA-494928 494928
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MSA-494928 Supplier MSA™ Supplier No. 494928
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Blow out kit 3000 PSI connects air supply hoses to manifold and air sources.

Specifications

Includes 26 to 30 ft./lb. torque, 0.75 in. socket, safety plug
Model 494928
Product Type Gasket Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.