Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ Custom Air V™ Respirator Accessories

Catalog No. 189993178 Shop All MSA Products
Change view
Click to view available options
For Use With (Equipment):
Custom Air V Escape Respirator
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
18-999-3178 Custom Air V Escape Respirator
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-999-3178 Supplier MSA™ Supplier No. 488899
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Color Yellow
For Use With (Equipment) Custom Air V Escape Respirator
Product Type Wall-mounted Case
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.