Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ Duo-Twin Pressure Demand Respirator Accessory, Breathing Tube Assembly

Catalog No. 18617 Shop All MSA Products
Change view
Click to view available options
For Use With (Equipment):
Duo-Twin Pressure Demand Respirator
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
18-617 Duo-Twin Pressure Demand Respirator
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-617 Supplier MSA™ Supplier No. 457158
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Breathing tube is designed for use with supplied air respirators.

  • For supplied air systems
  • Constant flow
  • Black
  • 28' Breathing Tube

Specifications

Color Black
Material Synthetic Rubber
Product Type Breathing Tube
For Use With (Equipment) Duo-Twin Pressure Demand Respirator
Model 457158
Length (English) 28 in.
Length (Metric) 71.12 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.