Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ Escape Respirator Accessory, Storage Case

Catalog No. 189993151 Shop All MSA Products
Change view
Click to view available options
Color:
Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
18-999-3151 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-999-3151 Supplier MSA™ Supplier No. 696192
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Resists weathering, moisture and corrosion to ensure escape respirators stay in ready to use condition.

  • Chemical resistant
  • Single wall mounting unit
  • Yellow color
  • Dimensions of 29.2 × 57.8 × 24.13cm (11.5 × 22.75 × 9.5 in.)

Specifications

Product Type Case
Color Yellow
Material ABS Plastic
Model 696192
For Use With (Equipment) Escape Respirator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.