Learn More
MSA™ ESP™ Full Facepiece Respirator Accessory, Mask Communications System
Catalog No. 189993213 Shop All MSA Products
Change view
Click to view available options
Amplifier Esp Ultv Scba
Catalog No.
18-999-3213 Amplifier Esp Ultv Scba
1 options
Catalog No. 18-999-3213 Supplier MSA™ Supplier No. 811737
  • Battery-powered self-contained communications unit for use with MSA full-facepiece respirators
  • Microphone and speaker housed in polymer case provide hands-free voice amplification for short-range communication
Order Info
Case is durable and waterproof but weighs only 8 oz. (0.23kg). Note:battery not included.
Specifications
Product Type Communications System
For Use With (Application) Chemical and nuclear industries and other environments where full facepiece air purifying respirator
For Use With (Equipment) ESP Full Facepiece Respirator
Model 811737
Weight (English) 8 oz.
Weight (Metric) 0.23 kg