Learn More
MSA™ Evolution™ 4000 Thermal Imaging Camera Battery
Catalog No. 19065197 Shop All MSA Products
Change view
Click to view available options
Product Type:
NiMH Battery
Catalog No. Product Type
19-065-197 NiMH Battery
1 options
Catalog No. 19-065-197 Supplier MSA™ Supplier No. 10016001
Specifications
Product Type NiMH Battery
For Use With (Equipment) Evolution™ 4000 thermal imaging camera
Voltage 7.2 V