Learn More
MSA™ Evolution™ 5200 Thermal Imaging Camera
Catalog No. 191304007 Shop All MSA Products
Change view
Click to view available options
Includes:
Charger, 1 Battery
Charger, Carrying Case, 2 Batteries
Charger, Quick-Temp/Heat Seeker PLUS option, Lanyard, Carrying Case, 2 Batteries, Reflective Kit
Charger, Quick-Temp/Heat Seeker PLUS option, Wrist Strap, Carrying Case, 2 batteries, Reflective Kit
Truck-mounted Charger, 2 Batteries
Truck-mounted Charger, Quick-Temp/Heat Seeker PLUS option, Lanyard, 2 Batteries
Truck-mounted Charger, Quick-Temp/Heat Seeker PLUS option, Wrist Strap, 2 Batteries
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
19-130-4007 Truck-mounted Charger, 2 Batteries
19-130-4002 Truck-mounted Charger, Quick-Temp/Heat Seeker PLUS option, Wrist Strap, 2 Batteries
19-130-4000 Charger, 1 Battery
19-130-4003 Charger, Quick-Temp/Heat Seeker PLUS option, Lanyard, Carrying Case, 2 Batteries, Reflective Kit
19-130-4001 Charger, Quick-Temp/Heat Seeker PLUS option, Wrist Strap, Carrying Case, 2 batteries, Reflective Kit
19-130-4004 Truck-mounted Charger, Quick-Temp/Heat Seeker PLUS option, Lanyard, 2 Batteries
19-130-4006 Charger, Carrying Case, 2 Batteries
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 19-130-4007 Supplier MSA™ Supplier No. 10063761
Delivers high-sense image quality up to 320°F, matching the temperature range at which most structural firefighting occurs
Camera offers a super-high Instantaneous Scene Dynamic Range† (ISDR) of 4795, providing clearer, sharper, more detailed images over the entire temperature range. RFI shielded unit is situated inside a high-impact housing with external rubber bumpers and internal mounting foam (which insulates sensor and electronics).
Performance Features
  • Super-wide High Sense operating range allows camera to stay in high sense mode longer
  • High sensitivity in Low Sense mode provides better image quality at higher scene temperatures
  • Increased ambient temperature tolerance permits unit to stay in high heat 60-100% longer
  • Indigo 160 x 120 microbolometer offers excellent image quality in a small format thermal imager
  • 30Hz frame rate reduces the choppiness associated with lower frame rates
  • Optional Heat Seeker PLUS color detail allows user to easily locate hot spots and the seat of the fire
Additional Features
  • Instant on with instant mode switching; 15% Low Sense activation
  • Warnings on all shut downs
  • Shutter indicator
  • Optional Quick-Temp digital temperature display with temperature scale measurement also available
†Instantaneous Scene Dynamic Range is the combined value of a sensor's sensitivity and temperature range to determine the number of temperature differences that can be seen at any given instant.
‡Also includes reflective kit.
Specifications
Product Type Thermal Imaging Camera
Includes Truck-mounted Charger, 2 Batteries
IP Rating IP67
Model Evolution™ 5200