Learn More
MSA™ Evolution™ 5200 Thermal Imaging Camera
Delivers high-sense image quality up to 320°F, matching the temperature range at which most structural firefighting occurs
Supplier:  MSA™ 10063768
 View more versions of this product
Catalog No. 19-130-4000
Description
Description
Camera offers a super-high Instantaneous Scene Dynamic Range† (ISDR) of 4795, providing clearer, sharper, more detailed images over the entire temperature range. RFI shielded unit is situated inside a high-impact housing with external rubber bumpers and internal mounting foam (which insulates sensor and electronics).
Performance Features
  • Super-wide High Sense operating range allows camera to stay in high sense mode longer
  • High sensitivity in Low Sense mode provides better image quality at higher scene temperatures
  • Increased ambient temperature tolerance permits unit to stay in high heat 60-100% longer
  • Indigo 160 x 120 microbolometer offers excellent image quality in a small format thermal imager
  • 30Hz frame rate reduces the choppiness associated with lower frame rates
  • Optional Heat Seeker PLUS color detail allows user to easily locate hot spots and the seat of the fire
Additional Features
  • Instant on with instant mode switching; 15% Low Sense activation
  • Warnings on all shut downs
  • Shutter indicator
  • Optional Quick-Temp digital temperature display with temperature scale measurement also available
Specifications
Specifications
Thermal Imaging Camera
Charger, 1 Battery
Evolution™ 5200
110/220 V
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.