Learn More
MSA™ Faceshield Assemblies
Catalog No. 189993160 Shop All MSA Products
Change view
Click to view available options
For Use With (Equipment):
Ultralite ii, Custom 4500 ii Air Mask
Catalog No. For Use With (Equipment)
18-999-3160 Ultralite ii, Custom 4500 ii Air Mask
1 options
This item is not returnable. View return policy
Catalog No. 18-999-3160 Supplier MSA™ Supplier No. 492006
Add to Cart
Add to Cart
This item is not returnable. View return policy
Specifications
Certifications/Compliance NIOSH 42CFR, Part 84 NIOSH CBRN NFPA 1981-2002, NFPA 1982-2002
Applications Firefighting, Confined Space
For Use With (Equipment) Ultralite ii, Custom 4500 ii Air Mask
Material Hycar Rubber
Includes Speed-on™ head hardness and nose cup
Size Medium
Product Type Faceshield Assembly