Learn More
MSA™ FireHawk™ M7 Air Mask Accessory, O-Ring
Catalog No. MSA630330 Shop All MSA Products
Catalog No. MSA630330 Supplier MSA™ Supplier No. 630330
Add to Cart
Add to Cart
The O-ring prevents leakage and is a critical component of the SCBA.
  • Replacement part for SCBA air system
  • Prevents air leakage
  • The O-ring should be inspected regularly and thoroughly checked based on usage
  • Found on the high pressure hose that connects to the air cylinder
Specifications
For Use With (Equipment) M7 Air Mask
Model 630330
Product Type O-ring