Learn More
MSA™ FireHawk™ M7 Air Mask Accessory, Silicone O-Ring
Catalog No. MSA630493 Shop All MSA Products
Change view
Click to view available options
Diameter (Metric):
2.82 cm
Catalog No. Diameter (Metric)
MSA630493 2.82 cm
1 options
Catalog No. MSA630493 Supplier MSA™ Supplier No. 630493
Add to Cart
Add to Cart
The O-ring prevents leakage and is a critical component of the SCBA.
  • Replacement part for SCBA air system
  • Prevents air leakage
  • The O-ring should be inspected regularly and thoroughly checked based on usage
  • Found on the high pressure hose that connects to the air cylinder
Specifications
For Use With (Equipment) M7 Air Mask
Model 630493
Diameter (Metric) 2.82 cm
Product Type O-ring