Learn More
MSA™ FireHawk™ M7 Air Masks with PASS
Catalog No. 17200010 Shop All MSA Products
Change view
Click to view available options
Low pressure, push-to-connect style regulator
Catalog No.
17-200-010 Low pressure, push-to-connect style regulator
1 options
This item is not returnable. View return policy
Catalog No. 17-200-010 Supplier MSA™ Supplier No. 10100091
This item is not returnable. View return policy
Includes
M-7 heads up display receiver, ipass
Ideal for industrial fire brigades
Designed to meet the needs of fire departments for basic, rugged and reliable SCBA without accessories.
  • Control module equipped with analog and digital pressure display for added redundancy
  • PASS icons display battery life, cylinder pressure, evacuation, thermal alarm and radio signal
  • Motion sensor alarm sounds at above 95dB
  • Datalogging of most recent 25 hours of use information
  • PR14™ first-stage regulator with 14 internal parts
  • Heads-up display and LED buddy lights
  • Stainless-steel cylinder band with secure cam-lock system
  • High-visibility reflective panels
  • Quick-Fill™ URC with Audi-Larm™ audible alarm
  • Firehawk™ MMR regulator
Order Info
Powered by four C cell batteries (not included). Facepiece and cylinder not included
Certifications
NIOSH 42 CFR Part 84 CBRN certified
Compliance
Meets NFPA 1981 and 1982-2007 Edition