Learn More
MSA™ FireHawk™ M7 Air Masks without PASS
Catalog No. 17200013 Shop All MSA Products
Change view
Click to view available options
Pressure:
High
Low
Model:
Push-to-connect with threaded hose and pressure gauge
Slide-to-connect with threaded hose and pressure gauge
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pressure Model
17-200-013 Low Slide-to-connect with threaded hose and pressure gauge
17-200-014 Low Push-to-connect with threaded hose and pressure gauge
17-200-015 High Slide-to-connect with threaded hose and pressure gauge
17-200-016 High Push-to-connect with threaded hose and pressure gauge
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 17-200-013 Supplier MSA™ Supplier No. 10121712
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Ideal for HazMat workers, first responders and firefighters
Basic, rugged and reliable SCBA without accessories.
  • Choice of intermittent or continuous mode; user can switch among HUD modes
  • Lower profile and easy-to-read pressure gauge
  • Can be used with existing standalone PASS devices
  • Upgrades easily to full PASS unit
  • Stainless-steel cylinder band with secure cam-lock system
  • High-visibility reflective panels
  • Bell alarm
Order Info
Facepiece and cylinder not included
Compliance
Meets NFPA 1981 Edition
Specifications
Certifications/Compliance NIOSH 42CFR, part 84 NIOSH CBRN NFPA 1981-2002, NFPA 1982-2002
Product Type Air Mask
Pressure Low
For Use With (Application) Ideal for hazmat workers, first responders, firefighters, confined space, firefighting, search and rescue
For Use With (Equipment) Standalone pass devices
Includes Threaded hose and pressure gauge, CBRN solid cover
Model Slide-to-connect with threaded hose and pressure gauge