Learn More
MSA™ FireHawk™ M7 Responder Air Mask
Catalog No. 12465428 Shop All MSA Products
Change view
Click to view available options
M7 Responder Air Mask
Catalog No.
12-465-428 M7 Responder Air Mask
1 options
This item is not returnable. View return policy
Catalog No. 12-465-428 Supplier MSA™ Supplier No. BM7HD14C0C12CABO
This item is not returnable. View return policy
Represents a true technological achievement in critical personal protection
  • Multipurpose responder mask available for CBRN self-contained breathing apparatus, air-purifying respiration and powered air-purifying respirator
  • Easily goes from first response and rescue to scene management and remediation
  • Provides the maximum level of protection when needed
  • One-way inhalation check valve to prevent cross-contamination
  • Wide-view lens with Kevlar SpeeD-ON™ head harness
  • High-performance mechanical speaking diaphragm
Specifications
Certifications/Compliance NIOSH 42CFR, part 84 NIOSH CBRN NFPA 1981-2002, NFPA 1982-2002
Product Type Facepiece
For Use With (Application) CBRN environments, confined space, clandestine labs, firefighting
For Use With (Equipment) CBRN, SCBA, APR, PAPR
Harness Style Dupont™ Kevlar™ aramid fiber Speed-on™